siRNA Details
| Gene Symbol: Gm9054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028005.1 | - | 3 | 237 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9054| Gene Symbol: Gm9054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028005.1 | - | 3 | 237 | ||