siRNA Details
| Gene Symbol: Ttc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133795.1 | NP_598556.1 | 2 | 370 | ||
| GenBank mRNA | AK019021.1 | - | 2 | 366 | ||
| AK077715.1 | - | 2 | 383 | |||
| AK166431.1 | - | 2 | 377 | |||
| AK170032.1 | - | 2 | 380 | |||
| AK170230.1 | - | 2 | 380 | |||
| BC010236.1 | - | 2 | 370 | 3581904 | ||

Ttc1
Genomic Map