siRNA Details
| Gene Symbol: Dnajc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024181.2 | NP_077143.2 | 15, 16 | 2041 | ||
| GenBank mRNA | AF255459.1 | - | 15, 16 | 1718 | ||
| AF314002.1 | - | 14, 15 | 1554 | |||
| AK004617.1 | - | 15, 16 | 1796 | |||
| AK028184.1 | - | 15, 16 | 1782 | |||
| AK128945.1 | - | 15, 16 | 1795 | |||
| AK165652.1 | - | 15, 16 | 1798 | |||
| AK166502.1 | - | 15, 16 | 1752 | |||
| AK169020.1 | - | 15, 16 | 1808 | |||
| BC002207.1 | - | 6, 7 | 603 | |||
| BC033461.1 | - | 15, 16 | 1780 | 4010341 | ||

Dnajc10
Genomic Map