siRNA Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025813.3 | NP_080089.1 | 14, 15 | 1387 | ||
| GenBank mRNA | AK004586.1 | - | 14, 15 | 1388 | ||
| AK032688.1 | - | 14, 15 | 1355 | |||
| AK075859.1 | - | 14, 15 | 1417 | |||
| AK135235.1 | - | 14, 15 | 1388 | |||
| AK136108.1 | - | 14, 15 | 1379 | |||
| AK152444.1 | - | 14, 15 | 1409 | |||
| AK159554.1 | - | 14, 15 | 1415 | |||
| AK166645.1 | - | 8, 9 | 779 | |||
| AK191801.1 | - | 4, 5 | 319 | |||
| AK214765.1 | - | 3, 4 | 185 | |||
| BC024891.1 | - | 14, 15 | 1369 | 3992247 | ||

Mfsd1
Genomic Map