siRNA Details
| Gene Symbol: Cpne8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033851.1 | NP_001029023.1 | 1, 2 | 229 | ||
| NM_025815.2 | NP_080091.1 | 1, 2 | 229 | |||
| GenBank mRNA | AK004559.1 | - | 1, 2 | 217 | ||
| AK005311.1 | - | 1, 2 | 163 | |||
| AK010227.1 | - | 1, 2 | 229 | |||
| AK013407.1 | - | 1, 2 | 214 | |||
| BC017626.1 | - | 1, 2 | 200 | 4502073 | ||
| BC048551.1 | - | 1, 2 | 229 | |||
| BC076564.1 | - | 1, 2 | 188 | |||
| BC138745.1 | - | 1, 2 | 228 | 8861767 | ||
| BC138746.1 | - | 1, 2 | 228 | 8861768 | ||

Cpne8
Genomic Map