siRNA Details
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024182.4 | NP_077144.2 | 8 | 921 | ||
GenBank mRNA | AK004748.1 | - | 8 | 1013 | ||
AK036631.1 | - | 1 | 1588 | |||
AK131635.1 | - | 8 | 925 | |||
AK134327.1 | - | 8 | 1013 | |||
AK147074.1 | - | 8 | 922 | |||
AK167309.1 | - | 8 | 1017 | |||
AK170882.1 | - | 8 | 1001 | |||
BC002255.1 | - | 5 | 451 | |||
BC033271.1 | - | 8 | 959 | 4016197 |
