siRNA Details
| Gene Symbol: Zfp442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177550.1 | NP_001171021.1 | 4 | 672 | ||
| GenBank mRNA | AB470967.1 | - | 4 | 679 | ||
| BC023805.1 | - | 4 | 671 | 5352880 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp442| Gene Symbol: Zfp442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177550.1 | NP_001171021.1 | 4 | 672 | ||
| GenBank mRNA | AB470967.1 | - | 4 | 679 | ||
| BC023805.1 | - | 4 | 671 | 5352880 | ||