siRNA Details
| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025834.3 | NP_080110.1 | 8 | 1021 | ||
| GenBank mRNA | AK005011.2 | - | 8 | 965 | ||
| AK008819.1 | - | 8 | 1022 | |||
| BC024104.1 | - | 8 | 1150 | 5136392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Proz| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025834.3 | NP_080110.1 | 8 | 1021 | ||
| GenBank mRNA | AK005011.2 | - | 8 | 965 | ||
| AK008819.1 | - | 8 | 1022 | |||
| BC024104.1 | - | 8 | 1150 | 5136392 | ||