siRNA Details
| Gene Symbol: Tmem107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025838.2 | NP_080114.1 | 4 | 406 | ||
| NM_028336.3 | NP_082612.2 | 4 | 406 | |||
| GenBank mRNA | AA217716.1 | - | 4 | 408 | ||
| AK002804.1 | - | 4 | 376 | |||
| AK003403.1 | - | 4 | 402 | |||
| AK012938.1 | - | 4 | 405 | |||
| AK154279.1 | - | 4 | 404 | |||
| BC096686.1 | - | 4 | 384 | 1446796 | ||
| BC120499.1 | - | 4 | 328 | 40129422 | ||
| BC137584.1 | - | 4 | 329 | 8860603 | ||
| CO044635.1 | - | 2 | 647 | |||

Tmem107
Genomic Map