siRNA Details
| Gene Symbol: Vps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF373710.1 | - | 10 | 772 | ||
| AK003572.2 | - | 10 | 802 | |||
| AK003699.1 | - | 10 | 726 | |||
| AK013852.1 | - | 10 | 723 | |||
| AK085669.1 | - | 7 | 1997 | |||
| BC013535.1 | - | 10 | 735 | 4239189 | ||

Vps28
Genomic Map