siRNA Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025857.2 | NP_080133.1 | 2 | 218 | ||
| GenBank mRNA | AK017714.1 | - | 2 | 187 | ||
| AK159897.1 | - | 2 | 219 | |||
| BC027311.1 | - | 2 | 214 | 3964905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aagab| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025857.2 | NP_080133.1 | 2 | 218 | ||
| GenBank mRNA | AK017714.1 | - | 2 | 187 | ||
| AK159897.1 | - | 2 | 219 | |||
| BC027311.1 | - | 2 | 214 | 3964905 | ||