siRNA Details
Gene Symbol: Cdca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025866.3 | NP_080142.1 | 2 | 187 | ||
GenBank mRNA | AK011289.1 | - | 2 | 188 | ||
AK028671.1 | - | 2 | 198 | |||
AK075882.1 | - | 2 | 152 | |||
AK153943.1 | - | 2 | 163 | |||
AK167734.1 | - | 2 | 188 | |||
BC066169.1 | - | 2 | 195 | 30474357 |
