siRNA Details
| Gene Symbol: Dusp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025869.3 | NP_080145.1 | 5 | 947 | ||
| GenBank mRNA | AB104416.1 | - | 3 | 737 | ||
| AK007513.1 | - | 3 | 317 | |||
| AK009781.1 | - | 5 | 948 | |||
| BC018204.1 | - | 3 | 737 | 4038120 | ||

Dusp26
Genomic Map