siRNA Details
| Gene Symbol: Luc7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025881.3 | NP_080157.1 | 4, 5 | 476 | ||
| NM_028190.3 | NP_082466.2 | 4, 5 | 476 | |||
| GenBank mRNA | AK010539.1 | - | 4, 5 | 428 | ||
| AK017655.1 | - | 4, 5 | 436 | |||
| AK160293.1 | - | 4, 5 | 477 | |||
| BC019194.1 | - | 5, 6 | 469 | |||
| BC034135.1 | - | 3, 4 | 642 | 5003664 | ||
| BC055875.1 | - | 4, 5 | 1459 | |||
| BQ572015.1 | - | 2, 3 | 128 | |||
| BY726224.1 | - | 5, 6 | 534 | |||

Luc7l
Genomic Map