siRNA Details
| Gene Symbol: Cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025892.2 | NP_080168.1 | 3 | 506 | ||
| GenBank mRNA | AK005324.1 | - | 3 | 506 | ||
| AK007713.1 | - | 3 | 490 | |||
| BC023715.1 | - | 2 | 186 | 5346460 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep19| Gene Symbol: Cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025892.2 | NP_080168.1 | 3 | 506 | ||
| GenBank mRNA | AK005324.1 | - | 3 | 506 | ||
| AK007713.1 | - | 3 | 490 | |||
| BC023715.1 | - | 2 | 186 | 5346460 | ||