siRNA Details
| Gene Symbol: Fam210b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC016210.1 | - | 3 | 527 | 4924329 | |
| BC057642.1 | - | 3 | 514 | 2655111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam210b| Gene Symbol: Fam210b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC016210.1 | - | 3 | 527 | 4924329 | |
| BC057642.1 | - | 3 | 514 | 2655111 | ||