siRNA Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023290.2 | NP_075779.2 | 5 | 761 | ||
| GenBank mRNA | AF277171.1 | - | 5 | 729 | ||
| AK009515.1 | - | 5 | 770 | |||
| AK011295.1 | - | 5 | 740 | |||
| AK077926.1 | - | 5 | 761 | |||
| BC025547.1 | - | 5 | 780 | 5320887 | ||

Mkrn2
Genomic Map