siRNA Details
| Gene Symbol: Upf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025924.2 | NP_080200.1 | 2, 3 | 381 | ||
| GenBank mRNA | AK011127.1 | - | 2, 3 | 358 | ||
| AK016053.1 | - | 2, 3 | 344 | |||
| AK020823.1 | - | 2, 3 | 385 | |||
| AK033239.1 | - | 2, 3 | 362 | |||
| AK145471.1 | - | 338 | ||||
| BC116988.1 | - | 2, 3 | 381 | 40126307 | ||
| BC119035.1 | - | 2, 3 | 381 | 8733784 | ||

Upf3a
Genomic Map