siRNA Details
| Gene Symbol: Rbm25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027349.3 | NP_081625.3 | 8, 9 | 986 | ||
| GenBank mRNA | AK011403.1 | - | 8, 9 | 967 | ||
| AK142132.1 | - | 2, 3 | 262 | |||
| AK151910.1 | - | 8, 9 | 938 | |||
| AK162135.1 | - | 1079 | ||||
| AK164367.1 | - | 8, 9 | 951 | |||
| BC010792.1 | - | 7, 8 | 732 | |||
| BC023749.1 | - | 6, 7 | 695 | |||
| BC158104.1 | - | 7, 8 | 866 | 9007388 | ||

Rbm25
Genomic Map