siRNA Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024188.6 | NP_077150.1 | 10 | 1362 | ||
| GenBank mRNA | AB085609.1 | - | 10 | 1104 | ||
| AK009027.1 | - | 10 | 1142 | |||
| AK011354.1 | - | 10 | 1042 | |||
| AK078086.1 | - | 10 | 1360 | |||
| AK146158.1 | - | 10 | 1120 | |||
| AK146342.1 | - | 10 | 1107 | |||
| AK151662.1 | - | 10 | 1101 | |||
| AK151695.1 | - | 10 | 1092 | |||
| AK167681.1 | - | 10 | 1192 | |||
| BC003422.1 | - | 10 | 1066 | 2598824 | ||

Oxct1
Genomic Map