siRNA Details
| Gene Symbol: Pus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023292.4 | NP_075781.3 | 4 | 1396 | ||
| GenBank mRNA | AF266505.1 | - | 3 | 1350 | ||
| AK030639.1 | - | 4 | 1792 | |||
| AK079095.1 | - | 4 | 1397 | |||
| BC029253.1 | - | 3 | 1357 | 4187198 | ||
| Gene Symbol: Hyls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083607.1 | - | 2 | 1940 | ||

Pus3
Genomic Map