siRNA Details
| Gene Symbol: Magt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190409.1 | NP_001177338.1 | 2 | 319 | ||
| NM_025952.4 | NP_080228.4 | 2 | 319 | |||
| GenBank mRNA | AK010320.1 | - | 2 | 255 | ||
| AK012185.1 | - | 2 | 196 | |||
| AK013243.1 | - | 2 | 200 | |||
| AK018623.1 | - | 2 | 222 | |||
| AK136622.1 | - | 2 | 201 | |||
| AK144696.1 | - | 2 | 198 | |||
| BC003881.1 | - | 2 | 206 | 3496572 | ||
| DQ000005.1 | - | 2 | 319 | |||

Magt1
Genomic Map