siRNA Details
| Gene Symbol: Tex40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039494.2 | NP_001034583.1 | 5 | 626 | ||
| GenBank mRNA | AK005644.1 | - | 5 | 605 | ||
| AK006034.1 | - | 5 | 564 | |||
| AK006134.1 | - | 5 | 629 | |||
| AK007089.1 | - | 5 | 602 | |||
| AK020779.1 | - | 1 | 393 | |||
| BC048452.1 | - | 5 | 563 | |||
| BC061075.1 | - | 5 | 568 | |||
| BC172038.1 | - | 4 | 814 | |||
| BC172041.1 | - | 4 | 869 | |||
| BC172042.1 | - | 4 | 783 | |||

Tex40
Genomic Map