siRNA Details
| Gene Symbol: 1700019D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144953.2 | NP_659202.1 | 2 | 583 | ||
| GenBank mRNA | AK006110.1 | - | 2 | 566 | ||
| AK006523.1 | - | 2 | 566 | |||
| AK010796.1 | - | 2 | 584 | |||
| AK010944.1 | - | 2 | 445 | |||
| BC048645.1 | - | 2 | 562 | 6772375 | ||

1700019D03Rik
Genomic Map