siRNA Details
| Gene Symbol: Timm21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025969.4 | NP_080245.1 | 2, 3 | 595 | ||
| GenBank mRNA | AK006594.1 | - | 2, 3 | 100 | ||
| AK012203.1 | - | 2, 3 | 595 | |||
| AK013281.1 | - | 2, 3 | 508 | |||
| AK017477.1 | - | 2, 3 | 563 | |||
| AK029037.1 | - | 2 | 675 | |||
| AK032468.1 | - | 2, 3 | 596 | |||
| AK168105.1 | - | 2, 3 | 506 | |||
| BC034297.1 | - | 2, 3 | 570 | 4511762 | ||

Timm21
Genomic Map