siRNA Details
| Gene Symbol: Zfp787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013012.1 | NP_001013030.1 | 2, 3 | 217 | ||
| GenBank mRNA | AK077458.1 | - | 2, 3 | 184 | ||
| AK159396.1 | - | 1, 2 | 42 | |||
| BC038658.1 | - | 2, 3 | 192 | |||
| BC137753.1 | - | 2, 3 | 139 | 8860772 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108904, XM_006228193.

Zfp787
Genomic Map