siRNA Details
Gene Symbol: Ndufa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025987.3 | NP_080263.1 | 2 | 255 | ||
GenBank mRNA | AK002749.1 | - | 2 | 202 | ||
AK010988.1 | - | 2 | 204 | |||
AK012328.1 | - | 2 | 204 | |||
BC048451.1 | - | 2 | 255 | 6773780 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC038336.1 | - | 295 | Chr. - - [-] on Build |
