siRNA Details
| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025992.2 | NP_080268.1 | 5 | 748 | ||
| GenBank mRNA | AK011047.1 | - | 5 | 635 | ||
| AK015214.1 | - | 790 | ||||
| AK017888.1 | - | 5 | 576 | |||
| AK149648.1 | - | 5 | 740 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Herc6| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025992.2 | NP_080268.1 | 5 | 748 | ||
| GenBank mRNA | AK011047.1 | - | 5 | 635 | ||
| AK015214.1 | - | 790 | ||||
| AK017888.1 | - | 5 | 576 | |||
| AK149648.1 | - | 5 | 740 | |||