siRNA Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025582.2 | NP_001020753.1 | 3 | 371 | ||
NM_001286986.1 | NP_001273915.1 | 3 | 368 | |||
NM_001286987.1 | NP_001273916.1 | 2 | 246 | |||
NM_026013.3 | NP_080289.1 | 3 | 371 | |||
GenBank mRNA | AI875295.1 | - | 3 | 371 | ||
AK008532.1 | - | 3 | 295 | |||
AK009940.1 | - | 3 | 308 | |||
AK012044.1 | - | 3 | 288 | |||
AK015888.1 | - | 3 | 334 | |||
AK052824.1 | - | 2 | 180 | |||
AK172455.1 | - | 3 | 338 | |||
BC024458.1 | - | 3 | 297 | 4975302 | ||
BI738450.1 | - | 3 | 339 |
