siRNA Details
| Gene Symbol: Zmat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026015.2 | NP_080291.1 | 2 | 149 | ||
| GenBank mRNA | AK003361.1 | - | 2 | 142 | ||
| AK007565.1 | - | 2 | 136 | |||
| AK012098.1 | - | 2 | 150 | |||
| AK013415.1 | - | 1 | 122 | |||
| BC031796.1 | - | 2 | 128 | 4205598 | ||

Zmat5
Genomic Map