siRNA Details
| Gene Symbol: Nde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114085.1 | NP_001107557.1 | 4 | 487 | ||
| NM_001285503.1 | NP_001272432.1 | 4 | 487 | |||
| NM_023317.2 | NP_075806.2 | 4 | 487 | |||
| GenBank mRNA | AF322073.1 | - | 3 | 394 | ||
| AK012830.1 | - | 4 | 417 | |||
| AK012861.1 | - | 4 | 437 | |||
| AK037289.1 | - | 4 | 430 | |||
| AK146982.1 | - | 4 | 417 | |||
| AK150826.1 | - | 4 | 477 | |||
| AK169743.1 | - | 4 | 417 | |||
| BC023267.1 | - | 4 | 466 | 3672565 | ||

Nde1
Genomic Map