siRNA Details
| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026032.1 | NP_080308.1 | 4 | 467 | ||
| GenBank mRNA | AK006006.1 | - | 2 | 202 | ||
| AK012814.1 | - | 2 | 202 | |||
| AK019398.1 | - | 2 | 197 | |||
| AK041290.1 | - | 4 | 467 | |||
| BC021460.1 | - | 4 | 750 | 5054997 | ||

Lsm1
Genomic Map