siRNA Details
| Gene Symbol: 2900011O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144518.3 | NP_653101.1 | 2 | 307 | ||
| GenBank mRNA | AK019307.1 | - | 2 | 300 | ||
| AK035868.1 | - | 2 | 284 | |||
| AK049997.1 | - | 2 | 339 | |||
| AK076136.1 | - | 2 | 326 | |||
| AK136859.1 | - | 2 | 307 | |||
| AK139470.1 | - | 2 | 319 | |||
| BC022741.1 | - | 2 | 286 | 4511454 | ||

2900011O08Rik
Genomic Map