siRNA Details
| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145456.2 | NP_663431.2 | 12, 13 | 2702 | ||
| GenBank mRNA | AK037945.1 | - | 8, 9 | 1297 | ||
| AK122528.1 | - | 11, 12 | 1970 | |||
| BC021311.1 | - | 3, 4 | 379 | |||
| BC021521.1 | - | 3, 4 | 582 | |||

Zswim6
Genomic Map