siRNA Details
| Gene Symbol: Med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026068.2 | NP_080344.2 | 3 | 788 | ||
| GenBank mRNA | AK005655.1 | - | 3 | 197 | ||
| AK013990.1 | - | 3 | 229 | |||
| AK045566.1 | - | 1 | 1266 | |||
| BC019685.1 | - | 3 | 258 | 3983617 | ||
| BC051929.1 | - | 3 | 788 | 5709831 | ||

Med31
Genomic Map