siRNA Details
| Gene Symbol: 3110040N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026077.3 | NP_080353.1 | 2, 3 | 216 | ||
| GenBank mRNA | AK014163.1 | - | 2, 3 | 196 | ||
| AK018003.1 | - | 2, 3 | 242 | |||
| AK019261.1 | - | 2, 3 | 216 | |||
| AK170168.1 | - | 2, 3 | 233 | |||
| BC027500.1 | - | 2, 3 | 204 | 1448876 | ||

3110040N11Rik
Genomic Map