siRNA Details
| Gene Symbol: Dock7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290636.1 | NP_001277565.1 | 20 | 2514 | ||
| NM_026082.5 | NP_080358.3 | 20 | 2514 | |||
| GenBank mRNA | BC094420.1 | - | 7 | 710 | ||
| CD802819.1 | - | 1 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock7| Gene Symbol: Dock7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290636.1 | NP_001277565.1 | 20 | 2514 | ||
| NM_026082.5 | NP_080358.3 | 20 | 2514 | |||
| GenBank mRNA | BC094420.1 | - | 7 | 710 | ||
| CD802819.1 | - | 1 | 166 | |||