siRNA Details
| Gene Symbol: Dock7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290636.1 | NP_001277565.1 | 21 | 2628 | ||
| NM_026082.5 | NP_080358.3 | 21 | 2628 | |||
| GenBank mRNA | BC086672.1 | - | 1 | 46 | 30647593 | |
| BC094420.1 | - | 8 | 824 | |||
| CD802819.1 | - | 2 | 280 | |||

Dock7
Genomic Map