siRNA Details
| Gene Symbol: Cltc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003908.1 | NP_001003908.1 | 12 | 2073 | ||
| GenBank mRNA | AK122193.1 | - | 12 | 2077 | ||
| BC035544.1 | - | 7 | 952 | |||
| BC079897.1 | - | 12 | 2073 | 30546407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cltc| Gene Symbol: Cltc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003908.1 | NP_001003908.1 | 12 | 2073 | ||
| GenBank mRNA | AK122193.1 | - | 12 | 2077 | ||
| BC035544.1 | - | 7 | 952 | |||
| BC079897.1 | - | 12 | 2073 | 30546407 | ||