siRNA Details
| Gene Symbol: Nanp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026086.2 | NP_080362.1 | 2 | 667 | ||
| GenBank mRNA | AK005566.1 | - | 2 | 643 | ||
| AK021344.1 | - | 2 | 654 | |||
| BC018527.1 | - | 2 | 638 | 3596532 | ||
| BC083086.1 | - | 2 | 667 | 6515732 | ||

Nanp
Genomic Map