siRNA Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038635.2 | NP_001033724.2 | 3 | 967 | ||
| NM_183262.3 | NP_899085.3 | 4 | 1717 | |||
| GenBank mRNA | AK006778.1 | - | 4 | 1482 | ||
| AK165988.1 | - | 3 | 862 | |||
| BC047277.1 | - | 3 | 1237 | |||
| BC138904.1 | - | 4 | 1624 | 8861926 | ||
| BC138905.1 | - | 4 | 1621 | 8861927 | ||

Stk35
Genomic Map