siRNA Details
| Gene Symbol: 1700121C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007220.1 | - | 148 | |||
| AK018972.1 | - | 148 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700121C10Rik| Gene Symbol: 1700121C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007220.1 | - | 148 | |||
| AK018972.1 | - | 148 | ||||