siRNA Details
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039951.2 | NP_001035040.1 | 4 | 607 | ||
| NM_026112.4 | NP_080388.2 | 4 | 607 | |||
| GenBank mRNA | AK032301.1 | - | 4 | 610 | ||
| AK039817.1 | - | 4 | 553 | |||
| AK044645.1 | - | 4 | 575 | |||
| AK139269.1 | - | 4 | 608 | |||
| AK143200.1 | - | 4 | 572 | |||
| AK165095.1 | - | 4 | 546 | |||
| AK173278.1 | - | 4 | 549 | |||
| BC052747.1 | - | 4 | 526 | 6837952 | ||
| BC110665.1 | - | 3 | 217 | |||

Zfp606
Genomic Map