siRNA Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207677.3 | NP_997560.3 | 2 | 207 | ||
| GenBank mRNA | AF457576.1 | - | 1 | 72 | ||
| AF543541.1 | - | 1 | 72 | |||
| AK135320.1 | - | 1 | 816 | |||
| BC092250.1 | - | 2 | 207 | 5043318 | ||
| CB589155.1 | - | 172 | ||||
| CX201206.1 | - | 1 | 59 | |||
| CX201977.1 | - | 2 | 204 | |||

Dedd2
Genomic Map