siRNA Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207677.3 | NP_997560.3 | 5 | 857 | ||
| GenBank mRNA | AF457576.1 | - | 4 | 722 | ||
| AF543541.1 | - | 4 | 722 | |||
| AK010701.1 | - | 5 | 669 | |||
| AK011713.1 | - | 4 | 563 | |||
| AK089096.1 | - | 3 | 229 | |||
| BC037043.1 | - | 4 | 483 | |||
| BC092250.1 | - | 5 | 857 | 5043318 | ||
| CX201206.1 | - | 4 | 710 | |||

Dedd2
Genomic Map