siRNA Details
| Gene Symbol: 1110008F13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026124.3 | NP_080400.1 | 1, 2 | 290 | ||
| GenBank mRNA | AK002452.1 | - | 1, 2 | 248 | ||
| AK003568.1 | - | 1, 2 | 285 | |||
| AK031002.1 | - | 1, 2 | 268 | |||
| AK179243.1 | - | 11 | ||||
| AK186989.1 | - | 11 | ||||
| BC019445.1 | - | 1, 2 | 228 | 5125733 | ||
| BY165552.1 | - | 1, 2 | 290 | |||

1110008F13Rik
Genomic Map