siRNA Details
| Gene Symbol: Cylc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026134.2 | NP_080410.2 | 5 | 1938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cylc1| Gene Symbol: Cylc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026134.2 | NP_080410.2 | 5 | 1938 | ||