siRNA Details
| Gene Symbol: Mfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024200.4 | NP_077162.2 | 6 | 788 | ||
| GenBank mRNA | AK018181.1 | - | 6 | 785 | ||
| AK141611.1 | - | 6 | 789 | |||
| AK187915.1 | - | 3 | 271 | |||
| AK220169.1 | - | 6 | 776 | |||
| AY174062.1 | - | 5 | 917 | |||
| BC008137.1 | - | 2 | 55 | |||
| BC056641.1 | - | 6 | 747 | 5097345 | ||

Mfn1
Genomic Map