siRNA Details
Gene Symbol: Ears2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014324.2 | - | 990 | |||
AK076043.1 | - | 5 | 984 | |||
AK129469.1 | - | 2325 | ||||
AK167506.1 | - | 5 | 1002 | |||
AK169225.1 | - | 5 | 991 | |||
BC025907.1 | - | 5 | 1008 | 5064949 |
