siRNA Details
| Gene Symbol: Adck3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163290.1 | NP_001156762.1 | 6 | 829 | ||
| NM_023341.3 | NP_075830.2 | 6 | 927 | |||
| GenBank mRNA | AJ278735.1 | - | 6 | 809 | ||
| AK004791.1 | - | 6 | 869 | |||
| AK014605.2 | - | 6 | 869 | |||
| AK178973.1 | - | 4 | 330 | |||
| AK184581.1 | - | 3 | 200 | |||
| AK187007.1 | - | 3 | 143 | |||
| AK207690.1 | - | 4 | 331 | |||
| BC030937.1 | - | 6 | 875 | 4922892 | ||
| U31629.1 | - | 4 | 468 | |||

Adck3
Genomic Map